Assay Details
| Gene Symbol: LOC101711512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845532.3 | XP_004845589.1 | 8 - 9 | 2250 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101711512| Gene Symbol: LOC101711512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845532.3 | XP_004845589.1 | 8 - 9 | 2250 | 98 | |
Set Membership: |
|