Assay Details
Gene Symbol: LOC101721350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848050.2 | XP_004848107.1 | 13 - 14 | 2355 | 89 | |
XM_021261051.1 | XP_021116710.1 | 13 - 14 | 2355 | 89 | ||
XM_021261052.1 | XP_021116711.1 | 13 - 14 | 2355 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
