Assay Details
Gene Symbol: LOC101721350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848050.2 | XP_004848107.1 | 1 - 2 | 704 | 72 | |
XM_021261051.1 | XP_021116710.1 | 1 - 2 | 704 | 72 | ||
XM_021261052.1 | XP_021116711.1 | 1 - 2 | 704 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004848050.2, XM_021261051.1, XM_021261052.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
