Assay Details
Gene Symbol: Ppm1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848055.2 | XP_004848112.1 | 2 - 3 | 789 | 71 | |
XM_004848056.2 | XP_004848113.1 | 3 - 4 | 866 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
