Assay Details
| Gene Symbol: Syt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855012.2 | XP_004855069.1 | 6 - 7 | 1587 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syt10| Gene Symbol: Syt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855012.2 | XP_004855069.1 | 6 - 7 | 1587 | 89 | |
Set Membership: |
|