Assay Details
Gene Symbol: Ipcef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856854.2 | XP_004856911.1 | 8 - 9 | 1155 | 81 | |
XM_004856855.3 | XP_004856912.1 | 8 - 9 | 1072 | 81 | ||
XM_004856856.3 | XP_004856913.1 | 8 - 9 | 1323 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
