Assay Details
Gene Symbol: Znf461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858998.2 | XP_004859055.1 | 4 - 5 | 866 | 64 | |
XM_004858999.2 | XP_004859056.1 | 4 - 5 | 869 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
