Assay Details
Gene Symbol: Tmbim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864601.2 | XP_004864658.1 | 10 - 11 | 892 | 59 | |
XM_004864602.2 | XP_004864659.1 | 9 - 10 | 803 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
