Assay Details
Gene Symbol: LOC101710773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864918.3 | XP_004864975.1 | 5 - 6 | 670 | 90 | |
XM_021244514.1 | XP_021100173.1 | 5 - 6 | 672 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
