Assay Details
| Gene Symbol: Fam111a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869727.2 | XP_004869784.1 | 3 - 4 | 1155 | 98 | |
| XM_013067500.2 | XP_012922954.1 | 2 - 3 | 520 | 98 | ||
| XM_021248293.1 | XP_021103952.1 | 2 - 3 | 211 | 98 | ||
| XM_021248294.1 | XP_021103953.1 | 2 - 3 | 816 | 98 | ||
| XM_021248295.1 | XP_021103954.1 | 3 - 4 | 1155 | 98 | ||
More Information
Set Membership: |
|

Fam111a
Genomic Map