Assay Details
| Gene Symbol: Map1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872855.3 | XP_004872912.1 | 5 - 6 | 7117 | 70 | |
| XM_021250683.1 | XP_021106342.1 | 4 - 5 | 6819 | 70 | ||
More Information
Set Membership: |
|

Map1b
Genomic Map