Assay Details
| Gene Symbol: Slc16a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873995.3 | XP_004874052.1 | 4 - 5 | 1062 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a10| Gene Symbol: Slc16a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873995.3 | XP_004874052.1 | 4 - 5 | 1062 | 81 | |
Set Membership: |
|