Assay Details
Gene Symbol: Ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013069340.2 | XP_012924794.1 | 3 - 4 | 426 | 64 | |
XM_013069341.2 | XP_012924795.1 | 3 - 4 | 412 | 64 | ||
XM_013069342.2 | XP_012924796.1 | 3 - 4 | 368 | 64 | ||
XM_021252303.1 | XP_021107962.1 | 3 - 4 | 372 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
