Assay Details
Gene Symbol: Mustn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021244160.1 | XP_021099819.1 | 2 - 3 | 166 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021244160.1 transcripts.
Set Membership: |
![]() ![]() |
