Assay Details
Gene Symbol: Mustn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021244160.1 | XP_021099819.1 | 3 - 4 | 304 | 106 | |
XM_021244161.1 | XP_021099820.1 | 3 - 4 | 294 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
