Assay Details
| Gene Symbol: Mfsd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004921227.3 | XP_004921284.2 | 3 - 4 | 564 | 63 | |
| XM_013069184.2 | XP_012924638.1 | 2 - 3 | 199 | 63 | ||
More Information
Set Membership: |
|

Mfsd4b
Genomic Map