Assay Details
| Gene Symbol: LOC101723434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863958.3 | XP_004864015.3 | 1 - 2 | 447 | 84 | |
| XM_004863959.3 | XP_004864016.2 | 1 - 2 | 481 | 84 | ||
More Information
Set Membership: |
|

LOC101723434
Genomic Map