Assay Details
| Gene Symbol: LOC101715954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834806.2 | XP_004834863.1 | 2 | 1113 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101715954| Gene Symbol: LOC101715954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834806.2 | XP_004834863.1 | 2 | 1113 | 93 | |
Set Membership: |
|