| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026179.hg.1 |
TC03001448.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026179.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026179.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026181.hg.1 |
TC03001448.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026181.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026181.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026177.hg.1 |
TC03001448.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026177.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026177.hg.1 |
ENST00000494383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026177.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013990.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013990.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013990.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013990.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013990.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013990.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013990.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013990.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026178.hg.1 |
TC03001448.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026178.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026178.hg.1 |
ENST00000494383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03026178.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207922.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300207922.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207922.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300207922.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207922.hg.1 |
TC0300013990.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300207924.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300207924.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207924.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300207924.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207924.hg.1 |
TC0300013990.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300207925.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300207925.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207925.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300207925.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207925.hg.1 |
TC0300013990.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013990.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013990.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013990.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013990.hg.1 |
ENST00000360658.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013990.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013990.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013990.hg.1 |
NM_017442.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013990.hg.1 |
NM_017442.3 |
Refseq |