Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320589.1 | NP_001307518.1 | 1 - 2 | 482 | 90 | |
| NM_001320590.1 | NP_001307519.1 | 1 - 2 | 482 | 90 | ||
| NM_138448.3 | NP_612457.1 | 1 - 2 | 482 | 90 | ||
| XM_017005410.1 | XP_016860899.1 | 1 - 2 | 352 | 90 | ||
| GenBank mRNA | BC012290.1 | - | 1 - 2 | 251 | 3864319 | 90 |
| BU857820.1 | - | 1 - 2 | 340 | 90 | ||
| X84195.1 | - | 1 - 2 | 231 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02004966.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02004762.hg.1 | TC02004966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004762.hg.1 | ENST00000303536.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004762.hg.1 | ENST00000394666.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004762.hg.1 | ENST00000406041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004762.hg.1 | ENST00000494922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004762.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004765.hg.1 | TC02004966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004765.hg.1 | ENST00000303536.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004765.hg.1 | ENST00000394666.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004765.hg.1 | ENST00000406041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004765.hg.1 | ENST00000494922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004765.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004759.hg.1 | TC02004966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004759.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004760.hg.1 | TC02004966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004760.hg.1 | ENST00000406041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004760.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004761.hg.1 | TC02004966.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004761.hg.1 | ENST00000303536.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004761.hg.1 | ENST00000394666.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004761.hg.1 | ENST00000406041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004761.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221794.hg.1 | TC0200016447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | ENST00000606865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016447.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221787.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221787.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221787.hg.1 | TC0200016447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221788.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221788.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221788.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221788.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221788.hg.1 | TC0200016447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | ENST00000494922.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221791.hg.1 | TC0200016447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221789.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221789.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221789.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221789.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221789.hg.1 | ENST00000303536.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221789.hg.1 | ENST00000394666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221789.hg.1 | ENST00000406041.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221789.hg.1 | NM_138448.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221789.hg.1 | TC0200016447.hg.1 | Clariom D TC |

ACYP2
Genomic Map