Assay Details
| Gene Symbol: DDT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001355.3 | NP_001346.1 | 1 - 2 | 104 | 141 | |
| GenBank mRNA | U49785.1 | - | 1 - 2 | 102 | 139 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001486.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001355.3,U49785.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010244.hg.1 | TC22001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010244.hg.1 | ENST00000350608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010244.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010238.hg.1 | TC22001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010238.hg.1 | ENST00000350608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010238.hg.1 | ENST00000398344.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010238.hg.1 | ENST00000403754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010238.hg.1 | ENST00000404092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010238.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010238.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010238.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010237.hg.1 | TC22001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010237.hg.1 | ENST00000350608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010237.hg.1 | ENST00000398344.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010237.hg.1 | ENST00000403754.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010237.hg.1 | ENST00000404092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010237.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010237.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010237.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147473.hg.1 | TC2200006874.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147473.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147473.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147473.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147473.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147473.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147473.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147473.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147473.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147466.hg.1 | TC2200006874.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147466.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147466.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147471.hg.1 | TC2200006874.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200147471.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147471.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147471.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147471.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147471.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200147471.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147471.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200147471.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | ENST00000215770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001084393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001144931.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001355.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006874.hg.1 | NM_001355.3 | Refseq |

DDT
Genomic Map