Assay Details
Gene Symbol: FABP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040442.1 | NP_001035532.1 | 3 - 4 | 283 | 82 | |
NM_001130958.1 | NP_001124430.1 | 4 - 5 | 363 | 82 | ||
NM_001445.2 | NP_001436.1 | 1 - 2 | 198 | 82 | ||
GenBank mRNA | AI208231.1 | - | 3 - 4 | 283 | 82 | |
DQ132786.1 | - | 4 - 5 | 364 | 82 | ||
U19869.1 | - | 1 - 2 | 119 | 82 | ||
X90908.1 | - | 1 - 2 | 198 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000893.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05012856.hg.1 | TC05000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05012856.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05012856.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05012859.hg.1 | TC05000893.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05012859.hg.1 | ENST00000393980.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05012859.hg.1 | ENST00000393982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05012859.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05012859.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05012859.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05012859.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009274.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500164813.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164813.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500164813.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164813.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500164813.hg.1 | TC0500009274.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | ENST00000393980.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | ENST00000402432.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | ENST00000521362.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | ENST00000523955.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | NM_001040442.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | NM_001130958.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | NM_001445.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500164817.hg.1 | TC0500009274.hg.1 | Clariom D TC |
