Assay Details
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001711.5 | NP_001702.1 | 1 - 2 | 223 | 100 | |
GenBank mRNA | AK222484.1 | - | 1 - 2 | 132 | 100 | |
AK223302.1 | - | 1 - 2 | 103 | 100 | ||
AK293268.1 | - | 1 - 2 | 159 | 100 | ||
AK297966.1 | - | 1 - 2 | 132 | 100 | ||
AU142113.1 | - | 1 - 2 | 159 | 100 | ||
DC419932.1 | - | 1 - 2 | 203 | 100 | ||
DC425011.1 | - | 1 - 2 | 184 | 100 | ||
J04599.1 | - | 1 - 2 | 107 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000750.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK293268.1,AK297966.1,J04599.1,NM_001711.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel remodeling
negative regulation of protein kinase activity
biological_process
cytokine-mediated signaling pathway
peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
extracellular matrix organization
glycosaminoglycan metabolic process
chondroitin sulfate biosynthetic process
chondroitin sulfate catabolic process
dermatan sulfate biosynthetic process
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
extracellular matrix structural constituent
glycosaminoglycan binding
extracellular matrix binding
blood vessel remodeling
negative regulation of protein kinase activity
biological_process
cytokine-mediated signaling pathway
peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
extracellular matrix organization
glycosaminoglycan metabolic process
chondroitin sulfate biosynthetic process
chondroitin sulfate catabolic process
dermatan sulfate biosynthetic process
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
extracellular matrix structural constituent
glycosaminoglycan binding
extracellular matrix binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X009738.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009738.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009738.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009738.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009732.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009732.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009733.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009733.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009733.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009740.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009740.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009740.hg.1 | ENST00000370204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009740.hg.1 | ENST00000430380.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009740.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009740.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009742.hg.1 | TC0X000750.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009742.hg.1 | ENST00000331595.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009742.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X009742.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159764.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159764.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159764.hg.1 | TC0X00008785.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00159766.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159766.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159766.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159766.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159766.hg.1 | TC0X00008785.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159771.hg.1 | TC0X00008785.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00159773.hg.1 | TC0X00008785.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000331595.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000431891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000472615.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000480756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | ENST00000492658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008785.hg.1 | NM_001711.5 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 171525 | - | - |
