Assay Details
| Gene Symbol: EFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001962.2 | NP_001953.1 | 1 - 2 | 408 | 98 | |
| XM_006714565.2 | XP_006714628.1 | 1 - 2 | 1877 | 98 | ||
| GenBank mRNA | AK308672.1 | - | 1 - 2 | 398 | 98 | |
| BC075054.2 | - | 1 - 2 | 175 | 30915365 | 98 | |
| BC075055.2 | - | 1 - 2 | 175 | 30915581 | 98 | |
| U26403.1 | - | 1 - 2 | 408 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001657.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
apoptotic process
activation of transmembrane receptor protein tyrosine kinase activity
nervous system development
axon guidance
regulation of cell-cell adhesion
regulation of cell morphogenesis
retinal ganglion cell axon guidance
regulation of actin cytoskeleton organization
regulation of GTPase activity
ephrin receptor signaling pathway
collateral sprouting
positive regulation of collateral sprouting
positive regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
regulation of focal adhesion assembly
positive regulation of synapse assembly
regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of microtubule cytoskeleton organization
negative regulation of substrate adhesion-dependent cell spreading
neurotrophin TRKA receptor binding
neurotrophin TRKB receptor binding
neurotrophin TRKC receptor binding
transmembrane receptor protein tyrosine kinase activator activity
chemorepellent activity
ephrin receptor binding
positive regulation of protein phosphorylation
apoptotic process
activation of transmembrane receptor protein tyrosine kinase activity
nervous system development
axon guidance
regulation of cell-cell adhesion
regulation of cell morphogenesis
retinal ganglion cell axon guidance
regulation of actin cytoskeleton organization
regulation of GTPase activity
ephrin receptor signaling pathway
collateral sprouting
positive regulation of collateral sprouting
positive regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
regulation of focal adhesion assembly
positive regulation of synapse assembly
regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of microtubule cytoskeleton organization
negative regulation of substrate adhesion-dependent cell spreading
neurotrophin TRKA receptor binding
neurotrophin TRKB receptor binding
neurotrophin TRKC receptor binding
transmembrane receptor protein tyrosine kinase activator activity
chemorepellent activity
ephrin receptor binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05022458.hg.1 | TC05001657.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022458.hg.1 | ENST00000333274.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022458.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000505499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000505499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022455.hg.1 | TC05001657.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022455.hg.1 | ENST00000333274.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022455.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022455.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022457.hg.1 | TC05001657.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022457.hg.1 | ENST00000333274.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022457.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022457.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022459.hg.1 | TC05001657.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022459.hg.1 | ENST00000333274.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022459.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180589.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180589.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180589.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180589.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180589.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180589.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180589.hg.1 | TC0500011648.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000505499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000505499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180583.hg.1 | TC0500011648.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500180590.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180590.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180590.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180590.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180590.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180590.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180590.hg.1 | TC0500011648.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500180587.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180587.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180587.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180587.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180587.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180587.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180587.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180587.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180587.hg.1 | TC0500011648.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000333274.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000504941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000505499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000505499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000509503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000510359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | ENST00000611503.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011648.hg.1 | NM_001962.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 158422 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 195013 | - | - |

EFNA5
Genomic Map