Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006185.hg.1 |
TC06000557.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006185.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006185.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155039.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155039.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155039.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155039.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155039.hg.1 |
TC0600007946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
ENST00000373256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007946.hg.1 |
NM_002062.4 |
Refseq |