Assay Details
| Gene Symbol: IDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289910.1 | NP_001276839.1 | 2 - 3 | 140 | 80 | |
| NM_002168.3 | NP_002159.2 | 2 - 3 | 373 | 80 | ||
| GenBank mRNA | AK222916.1 | - | 2 - 3 | 287 | 80 | |
| AK312627.1 | - | 2 - 3 | 337 | 80 | ||
| AK316388.1 | - | 2 - 3 | 214 | 80 | ||
| BC009244.2 | - | 2 - 3 | 275 | 2959540 | 80 | |
| BC071828.1 | - | 2 - 3 | 274 | 6090792 | 80 | |
| X69433.1 | - | 2 - 3 | 295 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178580.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178580.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178580.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178580.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178580.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178580.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178580.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178580.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178580.hg.1 | TC1500010422.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500178582.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178582.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178582.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178582.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178582.hg.1 | TC1500010422.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232153 | - | - |

IDH2
Genomic Map