Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005220.hg.1 |
ENST00000288309.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005220.hg.1 |
ENST00000400482.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005220.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005220.hg.1 |
TC21000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005223.hg.1 |
ENST00000400482.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005223.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005223.hg.1 |
TC21000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005222.hg.1 |
ENST00000400482.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005222.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005222.hg.1 |
TC21000443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005219.hg.1 |
ENST00000288309.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005219.hg.1 |
ENST00000400482.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005219.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005219.hg.1 |
TC21000443.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100154749.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154749.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154749.hg.1 |
TC2100008114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154749.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154749.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154752.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154752.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154752.hg.1 |
TC2100008114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154752.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154752.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008114.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154750.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154750.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154750.hg.1 |
TC2100008114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154750.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154750.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154751.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154751.hg.1 |
NM_002240.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154751.hg.1 |
TC2100008114.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154751.hg.1 |
ENST00000609713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154751.hg.1 |
NM_002240.4 |
Refseq |