Assay Details
Gene Symbol: MAGEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | X82539.1 | - | 3 - 4 | 265 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the X82539.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378981.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000378982.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397548.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | ENST00000397550.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002363.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_002367.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177404.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006878.hg.1 | NM_177415.2 | Refseq |
