Assay Details
| Gene Symbol: FAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000043.5 | NP_000034.1 | 2 - 3 | 512 | 85 | |
| NM_001320619.1 | NP_001307548.1 | 2 - 3 | 512 | 85 | ||
| NM_152871.3 | NP_690610.1 | 2 - 3 | 512 | 85 | ||
| NM_152872.3 | NP_690611.1 | 2 - 3 | 512 | 85 | ||
| NR_028033.3 | 2 - 3 | 512 | 85 | |||
| NR_028036.3 | 2 - 3 | 512 | 85 | |||
| NR_135313.1 | 2 - 3 | 512 | 85 | |||
| NR_135314.1 | 2 - 3 | 503 | 85 | |||
| XM_006717819.3 | XP_006717882.1 | 3 - 4 | 5194 | 85 | ||
| XM_011539764.2 | XP_011538066.1 | 3 - 4 | 583 | 85 | ||
| XM_011539765.2 | XP_011538067.1 | 3 - 4 | 371 | 85 | ||
| XM_011539766.2 | XP_011538068.1 | 3 - 4 | 323 | 85 | ||
| XM_011539767.2 | XP_011538069.1 | 2 - 3 | 246 | 85 | ||
| GenBank mRNA | AB209361.1 | - | 3 - 4 | 3829 | 85 | |
| AK290978.1 | - | 2 - 3 | 440 | 85 | ||
| AK311164.1 | - | 2 - 3 | 360 | 85 | ||
| AK311424.1 | - | 2 - 3 | 503 | 85 | ||
| AK311499.1 | - | 2 - 3 | 360 | 85 | ||
| AY495076.2 | - | 2 - 3 | 388 | 85 | ||
| BC012479.1 | - | 2 - 3 | 504 | 4514272 | 85 | |
| CR450307.1 | - | 2 - 3 | 194 | 85 | ||
| DA040126.1 | - | 2 - 3 | 371 | 85 | ||
| EU481974.2 | - | 2 - 3 | 388 | 85 | ||
| FJ200481.1 | - | 2 - 3 | 194 | 85 | ||
| FJ373045.1 | - | 2 - 3 | 194 | 85 | ||
| FJ373046.1 | - | 2 - 3 | 194 | 85 | ||
| FM246458.1 | - | 2 - 3 | 194 | 85 | ||
| FM246459.1 | - | 2 - 3 | 194 | 85 | ||
| M67454.1 | - | 2 - 3 | 388 | 85 | ||
| X63717.1 | - | 2 - 3 | 414 | 85 | ||
| X83492.1 | - | 2 - 3 | 219 | 85 | ||
| X83493.1 | - | 2 - 3 | 219 | 85 | ||
| X89101.1 | - | 2 - 3 | 194 | 85 | ||
| Z47993.1 | - | 2 - 3 | 194 | 85 | ||
| Z66556.1 | - | 2 - 3 | 194 | 85 | ||
| Z70519.1 | - | 2 - 3 | 194 | 85 | ||
| Z70520.1 | - | 2 - 3 | 194 | 85 | ||
More Information
Set Membership: |
|

FAS
Genomic Map