Assay Details
| Gene Symbol: NPY2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000910.3 | NP_000901.1 | 1 - 2 | 441 | 61 | |
| GenBank mRNA | U42766.1 | - | 1 - 2 | 447 | 61 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000782.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_000910.3,U42766.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
behavioral fear response
positive regulation of peptide secretion
outflow tract morphogenesis
cardiac left ventricle morphogenesis
cell surface receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
neuropeptide signaling pathway
nitric oxide mediated signal transduction
chemical synaptic transmission
locomotory behavior
feeding behavior
positive regulation of cell-substrate adhesion
negative regulation of cAMP biosynthetic process
negative regulation of neurological system process
positive regulation of dopamine secretion
positive regulation of smooth muscle contraction
positive regulation of circadian sleep/wake cycle, non-REM sleep
secretion
negative regulation of secretion
regulation of sensory perception of pain
negative regulation of synaptic transmission, glutamatergic
negative regulation of excitatory postsynaptic potential
negative regulation of feeding behavior
peptide YY receptor activity
receptor activity
neuropeptide Y receptor activity
calcium channel regulator activity
protein binding
behavioral fear response
positive regulation of peptide secretion
outflow tract morphogenesis
cardiac left ventricle morphogenesis
cell surface receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
neuropeptide signaling pathway
nitric oxide mediated signal transduction
chemical synaptic transmission
locomotory behavior
feeding behavior
positive regulation of cell-substrate adhesion
negative regulation of cAMP biosynthetic process
negative regulation of neurological system process
positive regulation of dopamine secretion
positive regulation of smooth muscle contraction
positive regulation of circadian sleep/wake cycle, non-REM sleep
secretion
negative regulation of secretion
regulation of sensory perception of pain
negative regulation of synaptic transmission, glutamatergic
negative regulation of excitatory postsynaptic potential
negative regulation of feeding behavior
peptide YY receptor activity
receptor activity
neuropeptide Y receptor activity
calcium channel regulator activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010780.hg.1 | TC04000782.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010780.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010780.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010782.hg.1 | TC04000782.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010782.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010782.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010782.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010781.hg.1 | TC04000782.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010781.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010781.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010781.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010786.hg.1 | TC04000782.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010786.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010786.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010786.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010785.hg.1 | TC04000782.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010785.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010785.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010785.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010788.hg.1 | TC04000782.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010788.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010788.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010788.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000508687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000508687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000508687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162328.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162328.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162328.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162328.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162328.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162328.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162328.hg.1 | TC0400009080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400162331.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162331.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162331.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162331.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162331.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162331.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162331.hg.1 | TC0400009080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400162334.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162334.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162334.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162334.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162334.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162334.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162334.hg.1 | TC0400009080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400162327.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162327.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162327.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162327.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162327.hg.1 | TC0400009080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009080.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162332.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162332.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162332.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162332.hg.1 | ENST00000329476.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162332.hg.1 | ENST00000506608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400162332.hg.1 | NM_000910.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400162332.hg.1 | TC0400009080.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000508687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000508687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000508687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012191.hg.1 | ENST00000511017.1 | Ensembl |

NPY2R
Genomic Map