Assay Details
| Gene Symbol: NCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000615.6 | NP_000606.3 | 18 - 19 | 2793 | 61 | |
| NM_001242607.1 | NP_001229536.1 | 20 - 21 | 2901 | 61 | ||
| NM_181351.4 | NP_851996.2 | 19 - 20 | 2823 | 61 | ||
| GenBank mRNA | AK292453.1 | - | 20 - 21 | 2458 | 61 | |
| AK299865.1 | - | 9 - 10 | 1004 | 61 | ||
| AK314589.1 | - | 20 - 21 | 2873 | 61 | ||
| BC014205.2 | - | 13 - 14 | 1687 | 61 | ||
| BC047244.1 | - | 19 - 20 | 2635 | 5590188 | 61 | |
| S71824.1 | - | 18 - 19 | 2628 | 61 | ||
| U63041.1 | - | 18 - 19 | 2464 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001015.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11012705.hg.1 | ENST00000316851.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012705.hg.1 | ENST00000397957.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012705.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012705.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012705.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012705.hg.1 | TC11001015.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012704.hg.1 | ENST00000316851.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012704.hg.1 | ENST00000397957.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012704.hg.1 | ENST00000531044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012704.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012704.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012704.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012704.hg.1 | TC11001015.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012703.hg.1 | ENST00000316851.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012703.hg.1 | ENST00000397957.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012703.hg.1 | ENST00000433634.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012703.hg.1 | ENST00000531044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012703.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012703.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012703.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11012703.hg.1 | TC11001015.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000528590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000531044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000611284.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000528590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000531044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000611284.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167284.hg.1 | TC1100009068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000528590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000531044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000611284.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000619839.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000528590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000531044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000611284.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | ENST00000619839.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167283.hg.1 | TC1100009068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167289.hg.1 | TC1100009068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000611284.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000611284.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167286.hg.1 | TC1100009068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000531044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000611284.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000531044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000611284.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167285.hg.1 | TC1100009068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000528158.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167287.hg.1 | TC1100009068.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000316851.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000533073.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000615112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000615285.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | ENST00000618266.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | NM_000615.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | NM_001242607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | NM_181351.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100167288.hg.1 | TC1100009068.hg.1 | Clariom D TC |

NCAM1
Genomic Map