Assay Details
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005261.3 | NP_005252.1 | 1 - 2 | 228 | 102 | |
XM_017013315.1 | XP_016868804.1 | 1 - 2 | 254 | 102 | ||
GenBank mRNA | DB018783.1 | - | 1 - 2 | 228 | 102 | |
U10550.1 | - | 1 - 2 | 202 | 102 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001432.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_005261.3,U10550.1,XM_017013315.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
immune response
mitotic nuclear division
signal transduction
cell surface receptor signaling pathway
small GTPase mediated signal transduction
chromosome organization
metaphase plate congression
magnesium ion binding
GTPase activity
protein binding
calmodulin binding
GTP binding
GDP binding
immune response
mitotic nuclear division
signal transduction
cell surface receptor signaling pathway
small GTPase mediated signal transduction
chromosome organization
metaphase plate congression
magnesium ion binding
GTPase activity
protein binding
calmodulin binding
GTP binding
GDP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08018582.hg.1 | TC08001432.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08018582.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08018582.hg.1 | ENST00000396194.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08018582.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08018582.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08018581.hg.1 | TC08001432.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08018581.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08018581.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08018579.hg.1 | TC08001432.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08018579.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08018579.hg.1 | ENST00000396194.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08018579.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08018579.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000523433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000523433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000523433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000523433.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175796.hg.1 | TC0800011132.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000523433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000523433.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011132.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175799.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175799.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175799.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175799.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175799.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175799.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175799.hg.1 | TC0800011132.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | ENST00000297596.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | ENST00000396194.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | ENST00000521817.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | ENST00000523660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | NM_005261.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | NM_181702.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800175800.hg.1 | TC0800011132.hg.1 | Clariom D TC |
