Assay Details
| Gene Symbol: PCDHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018905.2 | NP_061728.1 | 1 - 2 | 2495 | 88 | |
| GenBank mRNA | AF152310.1 | - | 1 - 2 | 2389 | 88 | |
More Information
Set Membership: |
|

PCDHA2
Genomic Map