| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013734.hg.1 |
TC09001045.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013734.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013730.hg.1 |
TC09001045.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013730.hg.1 |
ENST00000259607.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013730.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013730.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013733.hg.1 |
TC09001045.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013733.hg.1 |
ENST00000259607.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013733.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013731.hg.1 |
TC09001045.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013731.hg.1 |
ENST00000259607.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013731.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09013731.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168037.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168037.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168037.hg.1 |
TC0900009916.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009916.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168033.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168033.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168033.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168033.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168033.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168033.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168033.hg.1 |
TC0900009916.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900168036.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168036.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168036.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168036.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168036.hg.1 |
TC0900009916.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900168032.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168032.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168032.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168032.hg.1 |
ENST00000259607.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168032.hg.1 |
ENST00000378792.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900168032.hg.1 |
NM_002989.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900168032.hg.1 |
TC0900009916.hg.1 |
Clariom D TC |