| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014031.hg.1 |
TC06004106.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014031.hg.1 |
ENST00000341935.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014031.hg.1 |
ENST00000349984.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014031.hg.1 |
ENST00000400926.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014031.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014031.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014027.hg.1 |
TC06004106.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014027.hg.1 |
ENST00000341935.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014027.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014026.hg.1 |
TC06004106.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014026.hg.1 |
ENST00000341935.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014026.hg.1 |
ENST00000349984.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014026.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014029.hg.1 |
TC06004106.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014029.hg.1 |
ENST00000341935.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014029.hg.1 |
ENST00000349984.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06014029.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199929.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199929.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199929.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199929.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199929.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199929.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199929.hg.1 |
TC0600014204.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600199927.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199927.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199927.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199927.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199927.hg.1 |
TC0600014204.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600199926.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199926.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199926.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199926.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199926.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199926.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199926.hg.1 |
TC0600014204.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014204.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
ENST00000341935.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
ENST00000349984.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
ENST00000400926.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
NM_004367.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
NM_031409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199930.hg.1 |
TC0600014204.hg.1 |
Clariom D TC |