| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
ENST00000514165.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
ENST00000514165.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153615.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153615.hg.1 |
ENST00000514165.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153615.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153615.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153615.hg.1 |
ENST00000514165.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153615.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153615.hg.1 |
TC0500007834.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
ENST00000514165.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
ENST00000514165.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007834.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153616.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153616.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153616.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153616.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153616.hg.1 |
TC0500007834.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500153617.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153617.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153617.hg.1 |
ENST00000296677.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153617.hg.1 |
NM_005242.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153617.hg.1 |
TC0500007834.hg.1 |
Clariom D TC |