Assay Details
| Gene Symbol: CXCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000634.2 | NP_000625.1 | 1 - 2 | 86 | 72 | |
| GenBank mRNA | AK298647.1 | - | 1 - 2 | 95 | 72 | |
| AK309632.1 | - | 1 - 2 | 55 | 72 | ||
| AK312668.1 | - | 1 - 2 | 55 | 72 | ||
| BC028221.1 | - | 1 - 2 | 45 | 5217529 | 72 | |
| L19591.1 | - | 1 - 2 | 86 | 72 | ||
| U11871.1 | - | 1 - 2 | 67 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002767.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK298647.1,AK312668.1,BC028221.1,L19591.1,NM_000634.2,U11871.1 transcripts.
Set Membership: |
|

CXCR1
Genomic Map