Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000029.3 | NP_000020.1 | 1 - 2 | 508 | 71 | |
| GenBank mRNA | AK222798.1 | - | 1 - 2 | 39 | 71 | |
| AK222815.1 | - | 1 - 2 | 40 | 71 | ||
| AK293507.1 | - | 1 - 2 | 39 | 71 | ||
| AK303755.1 | - | 1 - 2 | 38 | 71 | ||
| AK307978.1 | - | 1 - 2 | 38 | 71 | ||
| AK312291.1 | - | 1 - 2 | 118 | 71 | ||
| AU128714.1 | - | 1 - 2 | 214 | 71 | ||
| DC412932.1 | - | 1 - 2 | 508 | 71 | ||
| K02215.1 | - | 1 - 2 | 39 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006281.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK293507.1,AK303755.1,AK312291.1,K02215.1,NM_000029.3 transcripts.
Set Membership: |
|

AGT
Genomic Map