Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037191.hg.1 |
TC03002094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037191.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037191.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037192.hg.1 |
TC03002094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037192.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037192.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037190.hg.1 |
TC03002094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037190.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03037190.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200642.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200642.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200642.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200642.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200642.hg.1 |
TC0300013469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300200638.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200638.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200638.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200638.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200638.hg.1 |
TC0300013469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300200639.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200639.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200639.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200639.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200639.hg.1 |
TC0300013469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300200640.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200640.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200640.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200640.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200640.hg.1 |
TC0300013469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300200641.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200641.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200641.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300200641.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300200641.hg.1 |
TC0300013469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
ENST00000287641.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013469.hg.1 |
NM_001048.3 |
Refseq |