Assay Details
| Gene Symbol: PTH2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309516.1 | NP_001296445.1 | 8 - 9 | 1233 | 66 | |
| NM_005048.3 | NP_005039.1 | 8 - 9 | 1129 | 66 | ||
| GenBank mRNA | AK294181.1 | - | 8 - 9 | 1233 | 66 | |
| AY497546.1 | - | 8 - 9 | 915 | 66 | ||
| BC036811.1 | - | 8 - 9 | 1128 | 5734330 | 66 | |
| U25128.1 | - | 8 - 9 | 1057 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001236.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018959.hg.1 | TC02001236.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018959.hg.1 | ENST00000272847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018959.hg.1 | ENST00000413482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018959.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175324.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175324.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175324.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175324.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175324.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175324.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175324.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175324.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175324.hg.1 | TC0200010627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 138697 | - | - |

PTH2R
Genomic Map