Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000462589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000462589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000462589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000474373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000474373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000474373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000583155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000583155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000583155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
ENST00000583155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025047.hg.1 |
TC01001622.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025047.hg.1 |
ENST00000367459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025047.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180791.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180791.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180791.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180791.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180791.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180791.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180791.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180791.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180791.hg.1 |
TC0100011012.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000367459.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000462589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000462589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000462589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000469578.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000474373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000474373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000474373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000498352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000583155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000583155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000583155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
ENST00000583155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011012.hg.1 |
NM_002922.3 |
Refseq |