Assay Details
Gene Symbol: CTSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004079.4 | NP_004070.3 | 3 - 4 | 510 | 82 | |
GenBank mRNA | AK226095.1 | - | 3 - 4 | 372 | 82 | |
AK314482.1 | - | 3 - 4 | 351 | 82 | ||
BC002642.2 | - | 3 - 4 | 311 | 3610589 | 82 | |
CR541676.1 | - | 2 - 3 | 249 | 82 | ||
M86553.1 | - | 2 - 3 | 255 | 82 | ||
M90696.1 | - | 3 - 4 | 385 | 82 | ||
S93414.1 | - | 2 - 3 | 255 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003210.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
toll-like receptor signaling pathway
adaptive immune response
proteolysis
immune response
response to acidic pH
protein processing
antigen processing and presentation
antigen processing and presentation of exogenous peptide antigen via MHC class II
extracellular matrix disassembly
collagen catabolic process
basement membrane disassembly
antigen processing and presentation of peptide antigen
proteolysis involved in cellular protein catabolic process
cellular response to thyroid hormone stimulus
positive regulation of cation channel activity
fibronectin binding
cysteine-type endopeptidase activity
serine-type endopeptidase activity
collagen binding
laminin binding
proteoglycan binding
toll-like receptor signaling pathway
adaptive immune response
proteolysis
immune response
response to acidic pH
protein processing
antigen processing and presentation
antigen processing and presentation of exogenous peptide antigen via MHC class II
extracellular matrix disassembly
collagen catabolic process
basement membrane disassembly
antigen processing and presentation of peptide antigen
proteolysis involved in cellular protein catabolic process
cellular response to thyroid hormone stimulus
positive regulation of cation channel activity
fibronectin binding
cysteine-type endopeptidase activity
serine-type endopeptidase activity
collagen binding
laminin binding
proteoglycan binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01049270.hg.1 | TC01003210.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01049270.hg.1 | ENST00000368985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01049270.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000607427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | ENST00000607427.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218014.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218014.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218014.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218014.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218014.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100218014.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100218014.hg.1 | TC0100015752.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000368985.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000448301.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000472977.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000480760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000483930.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000607427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | ENST00000607427.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_001199739.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015752.hg.1 | NM_004079.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 105790 | - | - |
