Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000377350.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000424945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000433890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000435926.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000470088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000489952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
NM_001307990.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
NM_004579.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000377350.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000424945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000433890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000435926.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000470088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
ENST00000489952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
NM_001307990.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
NM_004579.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204468.hg.1 |
TC1100013178.hg.1 |
Clariom D TC |