Assay Details
Gene Symbol: DEFB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004942.3 | NP_004933.1 | 1 - 2 | 204 | 81 | |
GenBank mRNA | BC069285.1 | - | 1 - 2 | 92 | 7262691 | 81 |
BC093983.1 | - | 1 - 2 | 79 | 7939828 | 81 | |
BC093985.1 | - | 1 - 2 | 83 | 7939830 | 81 | |
BM855542.1 | - | 1 - 2 | 120 | 81 | ||
HY264007.1 | - | 1 - 2 | 81 | |||
Z71389.1 | - | 1 - 2 | 79 | 81 |
Gene Symbol: DEFB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BE714509.1 | - | 1 - 2 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000051.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 4x180K | 102778 | AB064087.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 102778 | AB064088.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 102778 | AB064117.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 102778 | AM184037.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 45978 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 360335 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 74139 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 348276 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P157628 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P157628 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 35035 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 11061 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 45249 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P16375821 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P01857685 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P157628 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00332370 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P157628 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26219438 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16064714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26219438 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16064714 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P157628 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P38295854 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P01857685 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P05941594 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01857685 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P38295854 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P157628 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 6951 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 57204 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 11647 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 4328 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 94880 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 1950 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 79968 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16374678 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16375821 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05941594 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P16375821 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8149170 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8144486 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8144487 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8144483 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8144485 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8149171 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8144484 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8149169 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8144481 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 207356_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738511_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738511_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR08000471.hg.1 | TC08000051.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08000471.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08000471.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009463.hg.1 | ENST00000318157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009463.hg.1 | ENST00000318157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009463.hg.1 | ENST00000318157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009463.hg.1 | ENST00000318157.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009463.hg.1 | NM_001205266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009463.hg.1 | NM_001205266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009463.hg.1 | NM_001205266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08000472.hg.1 | TC08000051.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08000472.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08000472.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144998.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144998.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144998.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144998.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144998.hg.1 | TC0800006624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800145002.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800145002.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800145002.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800145002.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800145002.hg.1 | TC0800006624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800009463.hg.1 | ENST00000318157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009463.hg.1 | ENST00000318157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009463.hg.1 | ENST00000318157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009463.hg.1 | ENST00000318157.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009463.hg.1 | NM_001205266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009463.hg.1 | NM_001205266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009463.hg.1 | NM_001205266.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144997.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144997.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144997.hg.1 | TC0800006624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | ENST00000302247.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006624.hg.1 | NM_004942.3 | Refseq |
Human Genome U133 Plus 2.0 Array | 207356_at | - | - |
Human Genome U133 Plus 2.0 Array | 207356_PM_at | - | - |
Human Genome U133A 2.0 Array | 207356_PM_at | - | - |
Human Genome U133A 2.0 Array | 207356_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw515331 | - | - |
Human Genome U95 Set | 32464_at | - | - |
Human Genome U95 Set | 32464_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 207356_at3 | - | - |
Human Genome U133 Set | 207356_at | - | - |
Human Genome U133 Set | 207356_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036318 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036318 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2048043 | BE714509.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2048043 | NM_004942.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2048043 | BC069285.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2048043 | BC093983.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2048043 | BC093985.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680965 | NM_001205266.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680965 | BE714502.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680965 | NM_004942.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680965 | BC069285.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1680965 | BC093983.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2048043 | BE714509.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2048043 | NM_004942.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2048043 | BC069285.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2048043 | BC093983.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2048043 | BC093985.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2048043 | BE714509.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2048043 | NM_004942.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2048043 | BC069285.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2048043 | BC093983.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2048043 | BC093985.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2048043 | BE714509.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2048043 | NM_004942.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2048043 | BC069285.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2048043 | BC093983.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2048043 | BC093985.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_13124885-S | NM_004942.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_13124885-S | BC069285.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_13124885-S | BC093985.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_26250-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0006840594 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0006840594 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_26250 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_26250 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2048043 | BE714509.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2048043 | NM_004942.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2048043 | BC069285.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2048043 | BC093983.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2048043 | BC093985.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680965 | NM_001205266.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680965 | BE714502.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680965 | NM_004942.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680965 | BC069285.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1680965 | BC093983.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2048043 | BE714509.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2048043 | NM_004942.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2048043 | BC069285.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2048043 | BC093983.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2048043 | BC093985.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2048043 | BE714509.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2048043 | NM_004942.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2048043 | BC069285.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2048043 | BC093983.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2048043 | BC093985.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2048043 | BE714509.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2048043 | NM_004942.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2048043 | BC069285.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2048043 | BC093983.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2048043 | BC093985.1 | human_transcriptome |
