Assay Details
| Gene Symbol: WISP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003882.3 | NP_003873.1 | 3 - 4 | 717 | 81 | |
| GenBank mRNA | AF100779.1 | - | 3 - 4 | 699 | 81 | |
| AK293031.1 | - | 3 - 4 | 717 | 81 | ||
| AK315822.1 | - | 3 - 4 | 717 | 81 | ||
| BC074840.2 | - | 3 - 4 | 640 | 30915330 | 81 | |
| BC074841.2 | - | 3 - 4 | 640 | 30915551 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000770.hg.1
Set Membership: |
|

WISP1
Genomic Map