Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
TC09000746.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
ENST00000359428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
ENST00000411637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
ENST00000438605.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
ENST00000451030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
ENST00000465486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
ENST00000483497.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
ENST00000540899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
ENST00000541375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009558.hg.1 |
NM_005085.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000359428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000411637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000451030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000453861.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000465486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000483497.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000524578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000525384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000525980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000526346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000528114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000531929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
NM_005085.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000359428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000411637.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000451030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000453861.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000465486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000483497.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000524578.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000525384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000525980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000526346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000528114.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
ENST00000531929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
NM_005085.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161166.hg.1 |
TC0900008984.hg.1 |
Clariom D TC |