Assay Details
| Gene Symbol: AQP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318144.1 | NP_001305073.1 | 2 - 3 | 338 | 63 | |
| NM_004925.4 | NP_004916.1 | 2 - 3 | 338 | 63 | ||
| GenBank mRNA | AB001325.1 | - | 2 - 3 | 295 | 63 | |
| AJ493597.1 | - | 2 - 3 | 258 | 63 | ||
| AK292208.1 | - | 3 - 4 | 298 | 63 | ||
| AK303206.1 | - | 2 - 3 | 298 | 63 | ||
| AK315760.1 | - | 2 - 3 | 298 | 63 | ||
| BC013566.1 | - | 2 - 3 | 272 | 3877096 | 63 | |
| BT007199.1 | - | 2 - 3 | 235 | 63 | ||
| CR541991.1 | - | 3 - 4 | 235 | 63 | ||
| CR542025.1 | - | 3 - 4 | 235 | 63 | ||
| DA976040.1 | - | 2 - 3 | 338 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001012.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of immune system process
renal water homeostasis
transport
water transport
excretion
cellular water homeostasis
glycerol transport
response to retinoic acid
response to vitamin D
ion transmembrane transport
odontogenesis
regulation of keratinocyte differentiation
response to calcium ion
renal water absorption
urea transmembrane transport
transporter activity
urea transmembrane transporter activity
water channel activity
glycerol channel activity
positive regulation of immune system process
renal water homeostasis
transport
water transport
excretion
cellular water homeostasis
glycerol transport
response to retinoic acid
response to vitamin D
ion transmembrane transport
odontogenesis
regulation of keratinocyte differentiation
response to calcium ion
renal water absorption
urea transmembrane transport
transporter activity
urea transmembrane transporter activity
water channel activity
glycerol channel activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09013318.hg.1 | TC09001012.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013318.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013318.hg.1 | ENST00000343952.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013318.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013318.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013316.hg.1 | TC09001012.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013316.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013316.hg.1 | ENST00000343952.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013316.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013316.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013317.hg.1 | TC09001012.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013317.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013317.hg.1 | ENST00000343952.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013317.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013317.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013314.hg.1 | TC09001012.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013314.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013314.hg.1 | ENST00000343952.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013314.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013314.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013319.hg.1 | TC09001012.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013319.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013319.hg.1 | ENST00000343952.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013319.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013319.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167528.hg.1 | TC0900009860.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167525.hg.1 | TC0900009860.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900167530.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167530.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167530.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167530.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167530.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167530.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167530.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167530.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167530.hg.1 | TC0900009860.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167527.hg.1 | TC0900009860.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000297991.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000463983.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000473153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000493581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | ENST00000494313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009860.hg.1 | NM_004925.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 205859 | - | - |

AQP3
Genomic Map