Assay Details
| Gene Symbol: XPNPEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003399.5 | NP_003390.4 | 1 - 2 | 291 | 102 | |
| GenBank mRNA | AK300954.1 | - | 1 - 2 | 312 | 102 | |
| BC126174.1 | - | 1 - 2 | 208 | 8991890 | 102 | |
| BC143901.1 | - | 1 - 2 | 208 | 9052419 | 102 | |
| BP270314.1 | - | 1 - 2 | 282 | 102 | ||
| BX386512.2 | - | 1 - 2 | 291 | 102 | ||
| U90724.2 | - | 1 - 2 | 311 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000617.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK300954.1,BC126174.1,BC143901.1,NM_003399.5,U90724.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008099.hg.1 | TC0X000617.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008099.hg.1 | ENST00000371105.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008099.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008100.hg.1 | TC0X000617.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008100.hg.1 | ENST00000371105.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008100.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008100.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008103.hg.1 | TC0X000617.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008103.hg.1 | ENST00000371105.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008103.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008103.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008102.hg.1 | TC0X000617.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008102.hg.1 | ENST00000371105.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008102.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008102.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008101.hg.1 | TC0X000617.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008101.hg.1 | ENST00000371105.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008101.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008101.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157086.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157086.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157086.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157086.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157086.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157086.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157086.hg.1 | TC0X00008387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00157085.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157085.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157085.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157085.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157085.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157085.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157085.hg.1 | TC0X00008387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00157088.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157088.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157088.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157088.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157088.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157088.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157088.hg.1 | TC0X00008387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00157083.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157083.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157083.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157083.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157083.hg.1 | TC0X00008387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157084.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157084.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157084.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157084.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157084.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157084.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157084.hg.1 | TC0X00008387.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00157087.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157087.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157087.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157087.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157087.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157087.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157087.hg.1 | TC0X00008387.hg.1 | Clariom D TC |

XPNPEP2
Genomic Map