Assay Details
| Gene Symbol: ZFP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003408.2 | NP_003399.1 | 1 - 2 | 239 | 74 | |
| GenBank mRNA | AF022158.1 | - | 1 - 2 | 164 | 74 | |
| BC126390.1 | - | 1 - 2 | 207 | 8992106 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001496.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09018606.hg.1 | TC09001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018606.hg.1 | ENST00000374227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018606.hg.1 | ENST00000555206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018606.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018601.hg.1 | TC09001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018601.hg.1 | ENST00000374227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018601.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018601.hg.1 | ENST00000555206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018601.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018602.hg.1 | TC09001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018602.hg.1 | ENST00000374227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018602.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018602.hg.1 | ENST00000555206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018602.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018605.hg.1 | TC09001496.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018605.hg.1 | ENST00000374227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018605.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018605.hg.1 | ENST00000555206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018605.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176594.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176594.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176594.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176594.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176594.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176594.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176594.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176594.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176594.hg.1 | TC0900011247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176589.hg.1 | TC0900011247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900176595.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176595.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176595.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176595.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176595.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176595.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176595.hg.1 | TC0900011247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011247.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | ENST00000553380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176592.hg.1 | TC0900011247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | ENST00000374227.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | ENST00000555206.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | NM_001282515.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | NM_001282518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | NM_003408.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176593.hg.1 | TC0900011247.hg.1 | Clariom D TC |

ZFP37
Genomic Map