Assay Details
| Gene Symbol: DOC2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003585.4 | NP_003576.2 | 5 - 6 | 919 | 81 | |
| XM_011535363.2 | XP_011533665.1 | 4 - 5 | 735 | 81 | ||
| GenBank mRNA | D70830.1 | - | 5 - 6 | 928 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000961.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
vesicle fusion
protein localization
regulation of calcium ion-dependent exocytosis
positive regulation of vesicle fusion
positive regulation of insulin secretion
positive regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
calcium ion binding
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding
vesicle fusion
protein localization
regulation of calcium ion-dependent exocytosis
positive regulation of vesicle fusion
positive regulation of insulin secretion
positive regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
calcium ion binding
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 149227 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | chr17_random | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 53302 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3256396 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P236753 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P236753 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 31194 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P113404 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P113404 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15237 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 28655 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P12558599 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P20752584 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P12555139 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P12553855 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03199745 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P12558827 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3256396 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P236753 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3256396 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P236753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10185394 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30732928 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30732949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16932268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16932268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30732949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30732928 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10185394 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P236753 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P113404 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03809963 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P113404 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03809965 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P41920960 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03809959 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P236753 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 39216 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41920960 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P113404 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03809960 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41920951 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P21001273 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P413390 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P41920951 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P41920960 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P113404 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_003585_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8010902 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8010901 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 207311_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11737636_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11737636_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012705.hg.1 | ENST00000343572.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012705.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012705.hg.1 | TC17000961.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000609727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000609727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000609727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000609727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170804.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170804.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170804.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170804.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170804.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170804.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170804.hg.1 | TC1700009303.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000343572.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000609727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000609727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000609727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000609727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | ENST00000613549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009303.hg.1 | NM_003585.4 | Refseq |
| Human Genome U133 Plus 2.0 Array | 207311_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207311_at | - | - |
| Human Genome U133A 2.0 Array | 207311_at | - | - |
| Human Genome U133A 2.0 Array | 207311_PM_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003585_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw561481 | - | - |
| Human Genome U95 Set | 32422_at | - | - |
| Human Genome U95 Set | 32422_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207311_at4 | - | - |
| Human Genome U133 Set | 207311_PM_at | - | - |
| Human Genome U133 Set | 207311_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054959 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054959 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 121173 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1718401 | D70830.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1718401 | D70830.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1718401 | D70830.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1718401 | D70830.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1718401 | D70830.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_6005996-S | NM_003585.3 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_6005996-S | D70830.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_3915-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006370026 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006370026 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1718401 | D70830.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1718401 | D70830.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_3915 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_3915 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1718401 | D70830.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1718401 | D70830.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1718401 | D70830.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1718401 | D70830.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1718401 | NM_003585.3 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1718401 | D70830.1 | human_transcriptome |

DOC2B
Genomic Map